Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus B7-H7/HHLA2 Fc Chimera Protein
SDP

Catalog No. 10108B7050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10108B7050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10108B7050 Supplier R&D Systems™ Supplier No. 10108B7050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit anti-CD3 antibody induced IL-2 secretion in human T lymphocytes. The ED50 for this effect is 1-6 μg/mL.

Specifications

Accession Number XP_005548285
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Citric Acid and NaCl with Trehalose.
Gene ID (Entrez) 11148
Molecular Weight (g/mol) M.W. (Observed): 92-109 kDa, reducing conditions; M.W. (theoretical): 64 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey B7-H7/HHLA2 protein Cynomolgus Monkey B7-H7 (Ile21-Asn345) (N-terminus) Accession # XP_005548285 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Specific Reactivity B7-H7/HHLA2
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL aq. soln..
Buffer Lyophilized from a 0.2 μm filtered solution in Citric Acid and NaCl with Trehalose.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein B7-H7/HHLA2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.