Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus DNAM-1/CD226 Fc Chimera Protein

Catalog No. 9276DN100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-76D-N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 92-76D-N100 Supplier R&D Systems™ Supplier No. 9276DN100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey DNAM-1/CD226 Fc Chimera is immobilized at 0.25 μg/mL (100 μL/well), the concentration of Recombinant Human CD155/PVR Fc Chimera (Catalog # 9174-CD) that produces 50% of the optimal binding response is approximately 0.2-1 μg/mL.

Order Info

The product is shipped at ambient temperature.

Specifications

Accession Number XP_005586537
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 10666
Molecular Weight (g/mol) M.W. (Observed): 74-87 kDa, reducing conditions; M.W. (theoretical): 53 kDa
Name CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey DNAM-1/CD226 protein Cynomolgus Monkey DNAM-1/CD226 (Glu19-Asn247) (N-terminus) Accession # XP_005586537 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Biological Activity 0.2 to 1μg/mL (ED 50)
Product Type Recombinant protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Protein Tag No
Reconstitution Reconstitute at 1 mg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein DNAM-1/CD226
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.