Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus HGFR/c-MET Fc Chimera Protein

Catalog No. 11267ME100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11267ME100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11267ME100 Supplier R&D Systems™ Supplier No. 11267ME100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA. When Recombinant Human HGF (NS0-expressed) Protein (Catalog # 294-HGN ) is immobilized at 0.250 μg/mL (100 μL/well), Recombinant Cynomolgus Monkey HGFR/c-MET Fc Chimera (Catalog # 11267-ME) binds with an ED50 of 2.50-25.0 ng/mL.

Specifications

Accession Number EHH52447.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Molecular Weight (g/mol) M.W.-observed: 135-150 kDa, 40-45 kDa, and 103-115 kDa, under reducing conditions., M.W.-theroretical: 128 kDa, 33 kDa (alpha chain), and 96 kDa (beta chain)
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey HGFR/c-MET protein Cynomolgus HGFR/c-MET (Glu25-Thr932) (C-terminus) Accession # EHH52447.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.