Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus HVEM/TNFRSF14 Fc Chimera Protein
SDP

Catalog No. 9197HV100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-97H-V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 91-97H-V100 Supplier R&D Systems™ Supplier No. 9197HV100
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey HVEM/TNFRSF14 Fc Chimera is coated at 2 μg/mL (100 μL/well), biotinylated recombinant mouse BTLA Fc Chimera binds with a typical ED50 of 150-750 ng/mL.

Order Info

The product is shipped at ambient temperature.

Specifications

Accession Number XP_005545061
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8764
Molecular Weight (g/mol) M.W. (Observed): 56-63 kDa, reducing conditions; M.W. (theoretical): 44 kDa
Name ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 1
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey HVEM/TNFRSF14 protein Cynomolgus Monkey HVEM/TNFRSF14 (Pro37-Val203) (N-terminus) Accession # XP_005545061 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Biological Activity 50 to 750ng/mL (ED 50)
Product Type Recombinant protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Protein Tag No
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein HVEM/TNFRSF14
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.