Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus LILRB5/CD85c/LIR-8 His Protein
SDP

Catalog No. 10958T4050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10958T4050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10958T4050 Supplier R&D Systems™ Supplier No. 10958T4050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey LILRB5/CD85c/LIR-8 His-tag (Catalog # 10958-T4) is immobilized at 2 μg/mL (100 μL/well), Recombinant Human Angiopoietin-like Protein 7/ANGPTL7 (Catalog # 914-AN ) binds with an ED50 of 0.35-2.80 μg/mL.

Specifications

Accession Number XP_005590354.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 10990
Molecular Weight (g/mol) M.W.-observed: 62-70 kDa, reducing conditions., M.W.-theroretical: 48 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey LILRB5/CD85c/LIR-8 protein Gly24-Gly458, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD85 antigen-like family member C, CD85c, CD85c antigen, Leukocyte immunoglobulin-like receptor 8, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR8, LIR-8, LIR8CD85C, LIR-8 leukocyte immunoglobulin-like receptor subfamily B member 5, member 5
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein LIR-8/CD85c/LILRB5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.