Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Mesothelin (aa417-693) His Protein CF

Catalog No. 10764MS050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10764MS050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10764MS050 Supplier R&D Systems™ Supplier No. 10764MS050
Add to Cart
Add to Cart
This item is not returnable. View return policy

C-terminal

Measured by its binding ability in a functional ELISA. When Recombinant Human CA125/MUC16 (Catalog # 5609-MU ) is immobilized at 2.00 ug/mL (100 μL/well), Recombinant Cynomolgus Monkey Mesothelin C-Terminal (aa 417-693) His-tag protein binds with an ED50 of 0.700-4.20 μg/mL.

Specifications

Accession Number XP_015298164.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 10232
Molecular Weight (g/mol) M.W.-observed: 35-45 kDa, under reducing conditions, M.W.-theroretical: 32 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Mesothelin protein Asp417-Asp693, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein
Conjugate Unconjugated
Reconstitution Reconstitute at 1 mg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Mesothelin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.