Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey CD160 Fc Chimera Protein

Catalog No. 10197CD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-197-CD050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-197-CD050 Supplier R&D Systems™ Supplier No. 10197CD050
Add to Cart
Add to Cart
This item is not returnable. View return policy

CD160, also known as Natural killer cell receptor BY55, is a GPI-anchored member of the Ig superfamily that is expressed on both cytolytic lymphocytes and some unstimulated CD4+ T cells

  • Mature cynomolgus CD160 shares 91% aa sequence identity with human CD160
  • In human, it is expressed principally on non-myeloid hematopoietic cells
  • The effects of CD160 ligation appear to be context dependent

Specifications

Accession Number EHH50231
For Use With (Application) Bioactivity
Gene ID (Entrez) 11126
Molecular Weight (g/mol) 41 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey CD160 protein

Cynomolgus Monkey CD160 (Ile27-Ser159) Accession # EHH50231IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.