Learn More
Description
Order Info
Ships at ambient temperature
Specifications
Specifications
| Accession Number | EHH51019 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS |
| Gene ID (Entrez) | 4345 |
| Molecular Weight (g/mol) | 49 kDa |
| Name | CD200 Fc Chimera Recombinant Protein |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived cynomolgus monkey CD200 protein, (N-terminus) Cynomolgus Monkey CD200 (Gln56-Gly257) Accession &Num; EHH51019 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
