Learn More
R&D Systems™ Recombinant Cynomolgus Monkey CD43 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | EHH60309.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 6693 |
| Molecular Weight (g/mol) | M.W.-observed: 106-118 kDa, under reducing conditions., M.W.-theroretical: 50 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey CD43 protein Cynomolgus Monkey CD43 (Asp20-Thr258) (C-terminus) Accession # EHH60309.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD43, CD43 antigen, CD43), Galactoglycoprotein, GALGP, GPL115, Leukocyte sialoglycoprotein, Leukosialin, LSN, Ly-48, Sialophorin, sialophorin (gpL115, leukosialin, CD43), SPN |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.