Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey EpCAM/TROP1 Fc Protein

Catalog No. 10406EP050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-406-EP050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-406-EP050 Supplier R&D Systems™ Supplier No. 10406EP050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by the ability of the immobilized protein to support the adhesion of the L Cells mouse fibroblast cell line.

The ED50 for this effect is 0.3-1.8 μg/mL.

Specifications

Accession Number NP_001035118.1
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 4072
Molecular Weight (g/mol) Observed: 60-66 kDa, under reducing conditions; Theroretical: 54 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey EpCAM/TROP1 protein Cynomolgus Monkey EpCAM/TROP-1 (Gln24-Lys265) Accession # NP_001035118.1(N-terminus) IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Protein EpCAM/TROP1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.