Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey ErbB2/Her2 Fc Protein

Catalog No. 10526ER050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10526ER050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10526ER050 Supplier R&D Systems™ Supplier No. 10526ER050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured by its ability to block anti-ErbB2 mediated inhibition of SK-BR-3 human breast cancer cell proliferation. Brodowicz, T. et al. (1997) Int. J. Cancer 73:875. The ED50 for this effect is 0.2-1.2 μg/mL in the presence of 0.6 μg/mL Anti-Human ErbB2/Her2 Monoclonal Antibody (Trastuzumab, Catalog # MAB9589 ).

Specifications

Accession Number XP_005584091.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 2064
Molecular Weight (g/mol) M.W.-observed: 115-140 kDa, under reducing conditions, M.W.-theoretical: 96 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey ErbB2/Her2 protein Cynomolgus Monkey ErbB2/Her2 (Thr23-Thr652) (N-terminus) Accession # XP_005584091.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, Neu Oncogene, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGL, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, TKR1, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian)
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ErbB2/Her2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.