Learn More
R&D Systems™ Recombinant Cynomolgus Monkey Fc gamma RIIB Protein
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | NP_001271060 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and NaCl |
| Gene ID (Entrez) | 2213 |
| Molecular Weight (g/mol) | 21 to 35 kDa, reducing conditions |
| Name | CD32 antigen, CD32b, CD32c, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamm |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell,HEK293-derived cynomolgus monkey Fc gamma RIIB/CD32b protein Ala46-Pro217,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.