Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey GITR/TNFRSF18 Fc Chimera

Catalog No. 9428GR100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
94-28G-R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 94-28G-R100 Supplier R&D Systems™ Supplier No. 9428GR100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey GITR/TNFRSF18 Fc Chimera is immobilized at 0.5 μg/mL, 100 μL/well, the concentration of biotinylated recombinant human GITR Ligand/TNFSF18 that produces 50% of the optimal binding response is 8-40 ng/mL.

Order Info

Ships at ambient temperature.

Specifications

Accession Number XP_005545180
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 8784
Molecular Weight (g/mol) M.W. (Observed): 49-60 kDa, reducing conditions; M.W. (theoretical): 41 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey GITR/TNFRSF18 protein Cynomolgus Monkey GITR/TNFRSF18 (Gln20-Glu155) (N-terminus) Accession # XP_005545180 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Activation-inducible TNFR family receptor, AITR
Biological Activity 8 to 40ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein GITR/TNFRSF18
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.