Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey IgG1 Fc Protein
SDP

Catalog No. 9315HG100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-15H-G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-15H-G100 Supplier R&D Systems™ Supplier No. 9315HG100
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Cynomolgus Monkey IgG1 Fc Protein is derived from HEK293. The Recombinant Cynomolgus Monkey IgG1 Fc Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature

Specifications

Accession Number AAF14057
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 101867415
Molecular Weight (g/mol) 25 kDa
Name IgG1 Fc Chimera Recombinant Protein
Quantity 100 μg
Source Human embryonic kidney cell,HEK293-derived cynomolgus monkey IgG1 protein Pro110-Lys333
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Common Name IgG1
Biological Activity 0.2 to 1.2ng/mL
Product Type Recombinant Protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.