Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey IL-23R Fc Chimera Protein

Catalog No. 10306IR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-306-IR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-306-IR050 Supplier R&D Systems™ Supplier No. 10306IR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Human IL-23 Protein is immobilized at 1 μg/mL (100 μL/well), the concentration of Recombinant Cynomolgus Monkey IL-23R Fc Chimera that produces 50% of the optimal binding response is found to be approximately 50 to 250 ng/mL

Specifications

Accession Number XP_005543141
For Use With (Application) Bioactivity
Gene ID (Entrez) 149233
Molecular Weight (g/mol) 65 kDa
Name IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived cynomolgus monkey IL-23R protein

Cynomolgus Monkey IL-23R (Gly24-Asp353) Accession # XP_005543141IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Cynomolgus Monkey
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.