Learn More
R&D Systems™ Recombinant Cynomolgus Monkey IL-27 His-tag Protein
Description
Specifications
Specifications
| Accession Number | XP_005587614.1 (EBI-3) & XP_028697194.1 (IL-27a) |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with trehalose. |
| Gene ID (Entrez) | 246778 |
| Molecular Weight (g/mol) | Observed: 54-65 kDa, under reducing conditions; Theroretical: 50 kDa |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey IL-27 protein Cynomolgus Monkey EBI-3 (Arg36-Lys244) Accession # XP_005587614.1 GSGSSRGGSGSGGSGGGGSKL Cynomolgus Monkey IL-27 A *Unique sequence 6-His tag N-terminus C-terminus * Unique Sequence: This gene was isolated from a cynomolgus monkey cDNA library. The closest match to the obtained sequence is a genomic prediction for rhesus monkey, Accession XP_028697194.1 (Phe42-Pro254, A170T). |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 100 μg/mL in PBS. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.