Learn More
Description
Specifications
Specifications
| Accession Number | XP_005575320.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 10461 |
| Molecular Weight (g/mol) | M.W.-observed: 118-140 kDa, under reducing conditions, M.W.-theoretical: 79 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Mer protein Cynomolgus Monkey Mer (Ala23-Ala501) (N-terminus) Accession # XP_005575320.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
