Learn More
R&D Systems™ Recombinant Cynomolgus Monkey MMP-9 Protein
Description
Specifications
Specifications
| Accession Number | XP_005569271.2 |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, CaCl2, NaCl and Brij-35. |
| Gene ID (Entrez) | 4318 |
| Molecular Weight (g/mol) | M.W.-observed: 83-95 kDa, under reducing conditions, M.W.-theroretical: 76 kDa |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey MMP-9 protein Ala20-Asp707 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.