Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey TIGIT Fc Chimera Protein

Catalog No. 9380TG050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-80T-G050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-80T-G050 Supplier R&D Systems™ Supplier No. 9380TG050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human CD155/PVR (Catalog # 2530-CD ) is coated at 2.5 μg/mL (100 μL/well), the concentration of Recombinant Cynomolgus Monkey TIGIT Fc Chimera that produces 50% optimal binding response is 12-72 ng/mL.

Order Info

Ships at ambient temperature.

Specifications

Accession Number XP_005548158
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 201633
Molecular Weight (g/mol) M.W. (Observed): 43 - 60 kDa, reducing conditions; M.W. (theoretical): 40 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey TIGIT protein Cynomolgus Monkey TIGIT (Met22-Pro142) (N-terminus) Accession # XP_005548158 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Biological Activity 12 to 72ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein TIGIT
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.