Learn More
R&D Systems™ Recombinant Cynomolgus Monkey uPAR His-tag Protein
Description
Specifications
Specifications
| Accession Number | Q9GK78.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 5329 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 45-60 kDa, under reducing conditions.; MolecularWeight-theroretical: 32 kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived cynomolgus monkey uPAR protein Leu23-Arg303, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, PLAUR, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.