Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus OX40/TNFRSF4 Fc Chimera Protein

Catalog No. 10311OX050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-311-OX050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-311-OX050 Supplier R&D Systems™ Supplier No. 10311OX050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

Measured by its binding ability in a functional ELISA. When Recombinant Human OX40 Ligand/TNFSF4 (Catalog # 1054-OX ) is immobilized at 250 ng/mL, the concentration of Recombinant Cynomolgus Monkey OX40/TNFRSF4 Fc Chimera (Catalog# 10311-OX) that produces 50% optimal binding response is found to be approximately 5-30 ng/mL.

Specifications

Accession Number XP_005545179
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with trehalose.
Gene ID (Entrez) 7293
Molecular Weight (g/mol) M.W. (Observed): 62-72 kDa, under reducing conditions; M.W. (theoretical): 47 kDa
Name ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LOX40 antigen
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey OX40/TNFRSF4 protein Cynomolgus Monkey OX40/TNFRSF4 (Leu29-Ala216) (N-terminus) Accession # XP_005545179 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Cynomolgus Monkey
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein OX40/TNFRSF4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.