Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus PD-1 His-tag Protein

Catalog No. 8509PD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-09P-D050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-09P-D050 Supplier R&D Systems™ Supplier No. 8509PD050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human PD-L2/B7-DC Fc Chimera (Catalog # 1224-PL ) is coated at 0.5 μg/mL (100 μL/well), the concentration of Recombinant Cynomolgus Monkey PD-1, in the presence of Ms x Poly His, that produces 50% optimal binding response is typically 0.3-1.5 μg/mL.

Specifications

Accession Number NP_001271065.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in sterile PBS.
Gene ID (Entrez) 5133
Molecular Weight (g/mol) M.W. (Observed): 29-43 kDa, reducing conditions; M.W. (theoretical): 17 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey PD-1 protein Leu25-Gln167, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Common Name PD-1
Gene Symbol PDCD1
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human PD-L2/B7-DC Fc Chimera is coated at 0.5μg/mL (100μL/well), the concentration of Recombinant Cynomolgus Monkey PD-1, in the presence of Ms x Poly His, that produces 50% optimal binding response is typically 0.3-1.5μg/mL.
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Expression System Human embryonic kidney cell, HEK293-derived Leu25-Gln167, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.