Learn More
R&D Systems™ Recombinant Cynomolgus PD-L2/B7-DC Fc Chimera Protein
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | XP_005581839 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 80380 |
| Molecular Weight (g/mol) | M.W. (Observed): 62-77 kDa, reducing conditions; M.W. (theoretical): 49 kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived cynomolgus monkey PD-L2/B7-DC protein Cynomolgus Monkey PD-L2/B7-DC (Leu20-Pro219) (N-terminus) Accession # XP_005581839 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.