Learn More
R&D Systems™ Recombinant Cynomolgus/Rhesus FGL2 His-tag Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol with Trehalose. |
| Gene ID (Entrez) | 10875 |
| Molecular Weight (g/mol) | M.W.-observed: 54-78 kDa, under reducing conditions., M.W.-theroretical: 49 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived FGL2 protein Asn24-Pro439, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | fibrinogen-like 2, Fibrinogen-like protein 2, fibroleukin, pT49T49 |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.