Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus/Rhesus Macaque B7-2/CD86 Protein
SDP

Catalog No. 9798B2100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
97-98B-2100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 97-98B-2100 Supplier R&D Systems™ Supplier No. 9798B2100
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fc Chimera

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey CTLA-4 Fc Chimera (Catalog # 9336-CT ) is immobilized at 0.5 μg/mL (100 μL/well), Recombinant Cynomolgus Monkey/Rhesus Macaque B7-2/CD86 Fc Chimera (Catalog# 9798-B2) binds with an ED50 of 0.2-1 μg/mL.

Specifications

Accession Number XP_005548057
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS and NaCl with Trehalose.
Gene ID (Entrez) 942
Molecular Weight (g/mol) M.W. (Observed): 73-100 kDa, reducing conditions; M.W. (theoretical): 52 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived B7-2/CD86 protein Cynomolgus Monkey/Rhesus Macque B7-2 (Leu20-His239) (N-terminus) Accession # XP_005548057 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity B7-2/CD86
Conjugate Unconjugated
Species Cynomolgus Monkey, Rhesus macaque
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL aq. soln..
Buffer Lyophilized from a 0.2 μm filtered solution in MOPS and NaCl with Trehalose.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-2/CD86
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.