Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus/Rhesus NKG2D Fc Chimera Protein
SDP

Catalog No. 10960NK050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10960NK050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10960NK050 Supplier R&D Systems™ Supplier No. 10960NK050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey/Rhesus Macaque NKG2D/CD314 Fc Chimera (Catalog # 10960-NK) is immobilized at 2 μg/mL (100 μL/well), Biotinylated Recombinant Human MICA Fc Chimera Avi-tag (Catalog # AVI1300 ) binds with an ED50 of 25.0-200 ng/mL.

Specifications

Accession Number P61252.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 22914
Molecular Weight (g/mol) M.W.-observed: 45-65 kDa, under reducing conditions., M.W.-theroretical: 43 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived NKG2D/CD314 protein MD Human IgG1 (Pro100-Lys330) IEGR Cynomologus Monkey NKG2D (Trp74-Val216) (N-terminus) Accession # P61252.1 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein NKG2D/CD314
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.