Learn More
Creative Biomart Mecasermin; Recombinant human insulin-like growth factor-1 (rhIGF-1)
Supplier:  Creative Biomart THP0032
Encompass_Preferred
The product contains recombinant-DNA-engineered human insulin-like growth factor-1 (rhIGF-1). IGF-1 consists of 70 amino acids in a single chain with three intramolecular disulfide bridges and a molecular weight of 7649 daltons. The amino acid sequence of the product is identical to that of endogenous human IGF-1. The rhIGF-1 protein is synthesized in bacteria (E. coli) that have been modified by the addition of the gene for human IGF-1.
Catalog No. 50-208-4083