Learn More
Description
Specifications
Specifications
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 10mM HEPES buffer (pH7.4), 25mM NaCl |
| Molecular Weight (g/mol) | 14kDa |
| Name | GAGA-POZ domain Protein |
| Purification Method | Protein |
| Immunogen | MSLPMNSLYSLTWGDYGTSLVSAIQLLRCHGDLVDCTLAAGGRSFPAHKIVLCAASPFLLDLLKNTPCKHPVVMLAGVNANDLEALLEFVYRGEVSVDHAQLPSLLQAAQCLNIQGLAPQTVTKDDYTTH |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Drosophila |
| Research Category | Cell Cycle and Replication |
| Purity or Quality Grade | >95%, by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
