Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Drosophila Notch EGF20 Protein

Catalog No. 8117DN200
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8117DN200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8117DN200 Supplier R&D Systems™ Supplier No. 8117DN200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to be the acceptor substrate for Recombinant Mouse EOGT (Catalog # 8116-GT ). The specific activity of Recombinant Mouse EOGT is >0.7 pmol/min/ug using Notch Peptide as an acceptor and measured under the described conditions.

Specifications

Accession Number P07207
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 4851
Molecular Weight (g/mol) M.W. (Observed): 9 kDa, reducing conditions; M.W. (theoretical): 8.4 kDa
Name Notch EGF20
Quantity 200 μg
Source E. coli-derived drosophila Notch EGF20 protein Leader Sequence SPWPLD Notch (Asp791-Asn829) (N-terminus) Accession # P07207 Linker 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol N
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein Notch EGF20
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.