Learn More
Description
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 20% glycerol, 1mM DTT |
| Molecular Weight (g/mol) | TMW: 45.7kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Common Name | Acetate kinase |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
For research use only and is not approved for use in humans or in clinical diagnosis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
