Learn More
Abcam Recombinant E. coli beta Galactosidase protein (Tagged), 10UG
Supplier: Abcam AB26796710UG
Recombinant E. coli beta Galactosidase protein (Tagged), 10UG
Protein/ Peptide/ Enzyme Synonyms: b0344, JW0335, lacZ, Beta-galactosidase, Beta-gal, Lactase
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.