Learn More
Abcam Recombinant E. coli BirA protein (Active), 100UG
Supplier:  Abcam AB135015100UG
Encompass_Preferred
Recombinant E. coli BirA protein (Active) is a Escherichia coli K-12 Full Length protein, in the 2 to 321 aa range, expressed in Escherichia coli, with >=65% purity and suitable for SDS-PAGE, WB, FuncS.
Protein/ Peptide/ Enzyme Synonyms: bioR, dhbB, b3973, JW3941, birA, Bifunctional ligase/repressor BirA, Biotin operon repressor, Biotin--[acetyl-CoA-carboxylase] ligase, Biotin--protein ligase, Biotin-[acetyl-CoA carboxylase] synthetase
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3659
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.