Learn More
Description
Specifications
Specifications
| Accession Number | NP_416699 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 7.5), 2mM EDTA 10% glycerol |
| Gene ID (Entrez) | 949073 |
| Molecular Weight (g/mol) | 18.1kDa |
| Name | DsbE Protein |
| Purification Method | Affinity purified |
| Immunogen | MRNAEGDDPT NLESALIGKP VPKFRLESLD NPGQFYQADV LTQGKPVLLN VWATWCPTCR AEHQYLNQLS AQGIRVVGMN YKDDRQKAIS WLKELGNPYA LSLFDGDGML GLDLGVYGAP ETFLIDGNGI IRYRHAGDLN PRVWEEEIKP LWEKYSKEAA Q |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Gene Alias | ECK2187, yejQ |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
