Learn More
Abcam Recombinant E. coli fimH protein (His tag), 100UG
Supplier:  Abcam AB238375100UG
Encompass_Preferred
Recombinant E. coli fimH protein (His tag) is a Escherichia coli (strain K12) Full Length protein, in the 22 to 300 aa range, expressed in Yeast, with >90% purity and suitable for SDS-PAGE. The predicted molecular weight of ab238375 protein is 31.1 kDa. - Save time and ensure accurate results - use our E. coli Type 1 fimbrin D-mannose specific adhesin (FimH) protein as a control
Protein/ Peptide/ Enzyme Synonyms: b4320, JW4283, fimH, Type 1 fimbrin D-mannose specific adhesin, Protein FimH
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-6241
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.