Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant E. coli Glucose 6 Phosphate Dehydrogenase Protein
SDP

Catalog No. NBC119064 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC119064 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC119064 Supplier R&D Systems™ Supplier No. NBC119064

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids1-491 of E.coli Glucose 6 Phosphate Dehydrogenase The Recombinant E. coli Glucose 6 Phosphate Dehydrogenase Protein is derived from E. coli. The Recombinant E. coli Glucose 6 Phosphate Dehydrogenase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number NP_416366
For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol, 0.1mM PMSF, 2mM EDTA, 0.5mM DTT
Gene ID (Entrez) 2539
Molecular Weight (g/mol) 56kDa
Name Glucose 6 Phosphate Dehydrogenase Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Glucose 6 Phosphate Dehydrogenase,1-491aa, E.coli, Recombinant, E.coli (NP_416366).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Bacteria
Recombinant Recombinant
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.