Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant E. coli HDJ2 Protein
SDP

Catalog No. NBC118361 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118361 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118361 Supplier Novus Biologicals™ Supplier No. NBC118361
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-376 of E.coli HDJ2 The Recombinant E. coli HDJ2 Protein is derived from E. coli. The Recombinant E. coli HDJ2 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) ELISA, SDS-PAGE
Formulation 25mM Tris-HCl buffer (pH 8.8), 5mM DTT, 100mM NaCl, 10% glycerol
Gene ID (Entrez) 3301
Molecular Weight (g/mol) 41.1kDa
Name HDJ2 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 1-376aa
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.