Learn More
Description
Specifications
Specifications
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 25mM Tris-HCl buffer (pH7.5), 100mM NaCl, 5mM DTT, 10% glycerol |
| Gene ID (Entrez) | 3303 |
| Molecular Weight (g/mol) | 27.7kDa |
| Name | HSP70/HSPA1A Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Immunogen | MDVKDVLLLDVTPLSLGIETMGGVMTTLIAKNTTIPTKHSQVFSTAEDNQSAVTIHVLQGERKRAADNKSLGQFNLDGINPAPRGMPQIEVTFDIDADGILHVSAKDKNSGKEQKITIKASSGLNEDEIQKMVRDAEANAEADRKFEELVQTRNQGDHLLHSTRKQVEEAGDKLPADDKTAIESALTALETALKGEDKAAIEAKMQELAQVSQKLMEIAQQQHAQQQTAGADASANNAKDDDVVDAEFEEVKDKK |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Purity or Quality Grade | >95%, by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
