Learn More
Novus Biologicals™ Recombinant E. coli Nucleoside Diphosphate Kinase His Protein
Shop All R&D Systems Products

Specifications
Specifications
| Concentration | 1 mg/ml |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20 mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl, 1 mM DTT |
| Molecular Weight (g/mol) | M.W.-theoretical: 18 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | EC 2.7.4.6, Multifunctional nucleoside diphosphate kinase and apyrimidinic endonuclease and 3 '-phosphodiesterase, NDK, Nucleoside 5 '-diphosphate phosphotransferase, Nucleoside diphosphokinase |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Protein | Nucleoside Diphosphate Kinase |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.