Learn More
Novus Biologicals™ Recombinant E. coli Sec Protein Secretion Complex Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 10% glycerol |
| Molecular Weight (g/mol) | 17.2kDa |
| Name | Sec Protein Secretion Complex Protein |
| Purification Method | Protein |
| Immunogen | MSEQNNTEMT FQIQRIYTKD ISFEAPNAPH VFQKDWQPEV KLDLDTASSQ LADDVYEVVL RVTVTASLGE ETAFLCEVQQ GGIFSIAGIE GTQMAHCLGA YCPNILFPYA RECITSMVSR GTFPQLNLAP VNFDALFMNY LQQQAGEGTE EHQDA |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Bacteria |
| Purity or Quality Grade | >95%, by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.