Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant E. faecalis O-Glycosidase Protein

Catalog No. 8886GH020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-86G-H020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 88-86G-H020 Supplier R&D Systems™ Supplier No. 8886GH020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to hydrolyze p-Nitrophenyl galacto-N-bioside. The specific activity is >12,500 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q833X5
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Molecular Weight (g/mol) M.W. (Observed): 128 kDa, reducing conditions; M.W. (theoretical): 145 kDa
Quantity 20 μg
Source E. coli-derived e. faecalis O-Glycosidase protein Glu29-Lys1324, with N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Common Name O-Glycosidase
Biological Activity Measured by its ability to hydrolyze p-Nitrophenyl galacto-N-bioside. The specific activity is >12,500 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species E. faecalis
Recombinant Recombinant
Expression System E. coli-derived Glu29-Lys1324, with N-terminal Met and 6-His tag
Purity or Quality Grade >75%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein O-Glycosidase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.