Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant EBOV GP Protein

Catalog No. 9016EB100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-16E-B100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-16E-B100 Supplier R&D Systems™ Supplier No. 9016EB100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Zaire ebolavirus Glycoprotein

Measured by its binding ability in a functional ELISA. When Recombinant Viral EBOV GP is immobilized at 2 μg/mL (100 μL/well), the concentration of Recombinant Human CLEC10A/CD301 (Catalog # 4888-CL ) that produces 50% of the optimal binding response is approximately 20-100 ng/mL.

Specifications

Accession Number Q05320
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 3160774
Molecular Weight (g/mol) M.W. (Observed): 17-24 kDa and 105-120 kDa, reducing conditions; M.W. (theoretical): 69 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived viral EBOV GP protein Z. ebolavirus GP1 (Ile33-Arg501) (N-terminus) Accession # Q05320 Z. ebolavirus GP2 (Glu502-Gln650) (N-terminus) Accession # Q05320 HHHHHH (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Common Name EBOV GP
Gene Symbol GP
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Viral EBOV GP is immobilized at 2μg/mL (100μL/well), the concentration of Recombinant Human CLEC10A/CD301 that produces 50% of the optimal binding response is approximately 20-100ng/mL.
Conjugate Unconjugated
Species Virus
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL aq. soln..
Expression System Chinese Hamster Ovary cell line, CHO-derived Z. ebolavirus GP1 (Ile33-Arg501) Accession # Q05320, N-terminus; Z. ebolavirus GP2 (Glu502-Gln650) Accession # Q05320; HHHHHH, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EBOV GP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.