Learn More
Description
Specifications
Specifications
| Accession Number | P42160, P42160, P42160 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 2 x PBS, pH 7.4. |
| Gene ID (Entrez) | 3458 |
| Molecular Weight (g/mol) | 16.7 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of IFN-gamma as determined by LAL method. |
| Gene Alias | IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
