Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Equine PDGF-BB Protein
SDP

Catalog No. 8585BB010CF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8585BB010CF 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8585BB010CF Supplier R&D Systems™ Supplier No. 8585BB010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Equine PDGF-BB Protein is derived from E. coli. The Recombinant Equine PDGF-BB Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number XP_005606703
For Use With (Application) Bioactivity
Gene ID (Entrez) 5155
Molecular Weight (g/mol) Observed M.W.: 12kDa, reducing conditions, Theoretical M.W.: 12kDa
Quantity 10 μg
Source E. coli-derived equine PDGF-BB protein Ser51-Thr159
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name PDGF-BB
Biological Activity Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is typically 5-30ng/mL.
Conjugate Unconjugated
Species Equine
Recombinant Recombinant
Expression System E. coli-derived Ser51-Thr159
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.