Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Equine TGF-beta 1 Protein
SDP

Catalog No. 8617B1010CF Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8617B1010CF 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8617B1010CF Supplier R&D Systems™ Supplier No. 8617B1010/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Equine TGF-beta 1 Protein is derived from CHO. The Recombinant Equine TGF-beta 1 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_001075318
For Use With (Application) Bioactivity
Gene ID (Entrez) 7040
Molecular Weight (g/mol) Observed M.W.: 11-13kDa, reducing conditions, Theoretical M.W.: 13kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived equine TGF-beta 1 protein Ala279-Ser390 Accession &Num; NP_001075318
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Common Name TGF-beta 1
Gene Symbol TGFB1
Biological Activity Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED50 for this effect is typically 0.04-0.2ng/mL.
Conjugate Unconjugated
Species Equine
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ala279-Ser390 Accession # NP_001075318
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.