Learn More
Biomatik Corporation Recombinant Escherichia phage MS2 Capsid protein
Supplier:  Biomatik Corporation RPC26198
Encompass_Preferred
Recombinant Escherichia phage MS2 Capsid protein is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia phage MS2 (Bacteriophage MS2). Target Name: Escherichia phage MS2 Capsid protein. Target Synonyms: Coat protein. Accession Number: P03612. Expression Region: 2~130aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 21.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4625
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.