Learn More
R&D Systems™ Recombinant Escherichia T7 phage RNA polymerase Protein, CF
Shop All R&D Systems Products

Description
Specifications
Specifications
| Accession Number | P00573.2 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Phosphate, NaCl, TCEP and Glycerol. |
| Gene ID (Entrez) | 1261050 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 89-98 kDa, under reducing conditions, MolecularWeight-theroretical: 100 kDa |
| Quantity | 100 μg |
| Source | E. coli-derived e. coli T7 phage RNA polymerase protein Asn2-Ala883 with a N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | T7p07 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.