Learn More
Description
Specifications
Specifications
| Accession Number | WP_003034647.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, EDTA, Glycerol and TCEP. |
| Molecular Weight (g/mol) | Observed: 110-135 kDa, under reducing conditions |
| Quantity | 100 μg |
| Source | E. coli-derived f. tularensis Cpf1 protein APKKKRKVGIHGVPAA F. tularensis Cpf1 (Ser2-Asn1300) Accession # WP_003034647.1(N-terminus) KRPAATKKAGQAKKKKGG HHHHHH (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. |
| Protein | CRISPR-Cpf1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
