Learn More
Creative Biomart Turoctocog alfa, Recombinant factor VIII (rFVIII)
Supplier:  Creative Biomart THP0098
Encompass_Preferred
The product is a recombinant factor VIII (rFVIII) with a truncated B-domain made from the sequence coding for 10 amino acids from the N-terminus and 11 amino acids from the C-terminus of the naturally occurring B-domain. The product is produced in Chinese hamster ovary (CHO) cells without addition of any human- or animal-derived materials. During secretion, some rFVIII molecules are cleaved at the C-terminal of the heavy chain (HC) at amino acid 720, and a monoclonal antibody binding C-terminal to this position is used in the purification process allowing isolation of the intact rFVIII.
Catalog No. 50-208-4149
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.