Learn More
Creative Biomart Recombinant coagulation factor XIII A/F13A1, Catridecacog
Supplier:  Creative Biomart THP0062
Encompass_Preferred
The product, also known as catridecacog, is a recombinant form of the Factor XIII-A2 homodimer composed of two factor XIII (FXIII) A-subunits. Also known as Fibrin Stabilizing Factor (FSF), Factor XIII is an endogenously available coagulation factor and the final enzyme within the blood coagulation cascade. Within the body, FXIII circulates as a heterotetramer composed of 2 catalytic A-subunits and 2 non-catalytic B-subunits (FXIII-A2B2). When activated by thrombin at the site of injury, the FXIII A2B2 pro-enzyme is cleaved resulting in activation of the catalytic A-subunit and dissociation from its carrier B-subunit. When supplied as the recombinant form, the product binds to free human FXIII B-subunit resulting in a heterotetramer (rA2B2) with a similar activity profile and half-life as the endogenously available form.
Catalog No. 50-208-4113
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.