Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Feline IFN-gamma His Protein
SDP

Catalog No. NBP25950750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25950750 50 μg
NBP259507Y 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25950750 Supplier Novus Biologicals™ Supplier No. NBP25950750UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids24-167 of Feline IFN-gamma The Recombinant Feline IFN-gamma Protein is derived from E. coli. The Recombinant Feline IFN-gamma Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. PBS buffer (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 3458
Molecular Weight (g/mol) 19.3 kDa
Name Recombinant Feline IFN-gamma Protein
Purification Method >90%, by SDS-PAGE
Quantity 50 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Gene Symbol IFNG
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Feline
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.